Diba Omnifit® Frits for EZ, EZ SolventPlus™, BenchMark™, HiT™, and SolventPlus™ Columns
Select from a range of porosities for your media and chemistry
  • Extend the life of your columns with these replacement frits
Replacement frits are available in PTFE or PE and in a range of pore sizes, enabling you to select the most appropriate bed supports for your application. Match the column ID to select the correct frit.
4,900.00 - ₹38,700.00INR / Pkg of 20
40 variations of this product are available.
See All Product Options Below
40 PRODUCT OPTIONS
Compareitem specifications
Column Inside Diameter (ID) (mm) Wetted Materials Description
Availability Pricing
Diba Chromatography Column Frit, 5 mm dia, 5 µm, PTFE; 20/pk WW-11944-89 Mfr # 006FR-05-05
COMPARE
5 PTFE 5 µm
₹6,000.00INR / Pkg of 20
Diba Chromatography Column Frit, 5 mm dia, 10 µm, PTFE; 20/pk WW-11944-90 Mfr # 006FR-05-10
COMPARE
5 PTFE 10 µm
₹6,000.00INR / Pkg of 20
Diba Chromatography Column Frit, 5 mm dia, 20 µm, PTFE; 20/pk WW-11944-91 Mfr # 006FR-05-20
COMPARE
5 PTFE 20 µm
₹6,000.00INR / Pkg of 20
Diba Chromatography Column Frit, 5 mm dia, 25 µm, PE; 20/pk WW-11944-92 Mfr # 006FR-05-25-PE
COMPARE
5 PE 25 µm
₹6,000.00INR / Pkg of 20
Diba Chromatography Column Frit, 5 mm dia, 30 µm, PTFE; 20/pk WW-11944-93 Mfr # 006FR-05-30
COMPARE
5 PTFE 30 µm
₹6,000.00INR / Pkg of 20
Diba Chromatography Column Frit, 5 mm dia, 50 µm, PTFE; 20/pk WW-11944-94 Mfr # 006FR-05-50
COMPARE
5 PTFE 50 µm
₹6,000.00INR / Pkg of 20
Diba Chromatography Column Frit, 6.6 mm dia, 5 µm, PTFE; 20/pk WW-11944-95 Mfr # 006FR-06-05
COMPARE
6.6 PTFE 5 µm
₹4,900.00INR / Pkg of 20
Diba Chromatography Column Frit, 6.6 mm dia, 10 µm, PTFE; 20/pk WW-11944-96 Mfr # 006FR-06-10
COMPARE
6.6 PTFE 10 µm
₹4,900.00INR / Pkg of 20
Diba Chromatography Column Frit, 6.6 mm dia, 20 µm, PTFE; 20/pk WW-11944-97 Mfr # 006FR-06-20
COMPARE
6.6 PTFE 20 µm
₹4,900.00INR / Pkg of 20
Diba Chromatography Column Frit, 6.6 mm dia, 25 µm, PE; 20/pk WW-11944-98 Mfr # 006FR-06-25-PE
COMPARE
6.6 PE 25 µm
₹4,900.00INR / Pkg of 20
Diba Chromatography Column Frit, 6.6 mm dia, 30 µm, PTFE; 20/pk WW-11944-99 Mfr # 006FR-06-30
COMPARE
6.6 PTFE 30 µm
₹4,900.00INR / Pkg of 20
Diba Chromatography Column Frit, 6.6 mm dia, 50 µm, PTFE; 20/pk WW-11945-00 Mfr # 006FR-06-50
COMPARE
6.6 PTFE 50 µm
₹4,900.00INR / Pkg of 20
Diba Chromatography Column Frit, 10 mm dia, 5 µm, PTFE; 20/pk WW-11945-01 Mfr # 006FR-10-05
COMPARE
10 PTFE 5 µm
₹6,000.00INR / Pkg of 20
Diba Chromatography Column Frit, 10 mm dia, 10 µm, PTFE; 20/pk WW-11945-02 Mfr # 006FR-10-10
COMPARE
10 PTFE 10 µm
₹6,000.00INR / Pkg of 20
Diba Chromatography Column Frit, 10 mm dia, 20 µm, PTFE; 20/pk WW-11945-03 Mfr # 006FR-10-20
COMPARE
10 PTFE 20 µm
₹6,000.00INR / Pkg of 20
Diba Chromatography Column Frit, 10 mm dia, 25 µm, PE; 20/pk WW-11945-04 Mfr # 006FR-10-25-PE
COMPARE
10 PE 25 µm
₹6,000.00INR / Pkg of 20
Diba Chromatography Column Frit, 10 mm dia, 30 µm, PTFE; 20/pk WW-11945-05 Mfr # 006FR-10-30
COMPARE
10 PTFE 30 µm
₹6,000.00INR / Pkg of 20
Diba Chromatography Column Frit, 10 mm dia, 50 µm, PTFE; 20/pk WW-11945-06 Mfr # 006FR-10-50
COMPARE
10 PTFE 50 µm
₹6,000.00INR / Pkg of 20
Diba Chromatography Column Frit, 15 mm dia, 5 µm, PTFE; 20/pk WW-11945-07 Mfr # 006FR-15-05
COMPARE
15 PTFE 5 µm
₹13,300.00INR / Pkg of 20
Diba Chromatography Column Frit, 15 mm dia, 10 µm, PTFE; 20/pk WW-11945-08 Mfr # 006FR-15-10
COMPARE
15 PTFE 10 µm
₹13,300.00INR / Pkg of 20
Showing 1 - 20 of 40