Ohaus Pioneer Analytical Balances 62704
Draft shield removes completely for complete cleaning of all components within seconds
  • Easy to clean analytical draft shield – all panels remove easily for cleaning
  • Selectable environmental settings – three filter modes and adjustable zero tracking adjust balance sensitivity to environmental disturbances, or application requirements
Periodic cleaning helps avoid sample contamination and extends the life of the balance. A standard in-use cover provides added protection against spillage and minimizes damage to the display and keypad. Three user-selectable environmental settings help the balance adapt to harsh environments and other disturbances for greater weighing accuracy. An up front level indicator eliminates the need to look behind the balance during the leveling process -- a quick glance at the up-front indicator helps ensure the balance is level prior to each use.
157,000.00 - ₹187,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
Ohaus PA 64 Pioneer Series Analytical Balance, 65g x 0.1 Mg (230 V) WW-11009-46 Mfr # PA 64
COMPARE
External 65 0.0001
₹157,000.00INR / Each
Ohaus PA 114 Pioneer Series Analytical Balance, 110g x 0.1 Mg (230 V) WW-11009-47 Mfr # PA 114
COMPARE
External 110 0.0001
₹175,000.00INR / Each
Ohaus PA 214 Pioneer Series Analytical Balance, 210g x 0.1 Mg (230 V) WW-11009-48 Mfr # PA 214
COMPARE
External 210 0.0001
₹187,000.00INR / Each