Ohaus® Navigator™ Portable Balances with Touchless Sensor
Wide range of weighing applications in a rugged, compact design
  • Overload protection withstands loads up to four times the rated capacity
  • Integrated shipping lock protects weighing cell during transportation
  • Multiple application modes designed for versatility
  • Touchless sensor provides hands-free operation
Ohaus Navigator portable balances offer a wide range of use in industrial, food, and laboratory weighing applications. The balances stabilize quickly to provide fast, accurate, and repeatable weighing results—helps improve efficiency and increase productivity. A bright backlit LCD provides optimum visibility in any lighting condition, with large digits and indicators that make it fast and easy to read the weighing results. Touchless sensor allows you to zero out, print, tare, or change functions without touching the balance.
These multipurpose, portable balances feature a rugged ABS housing, stainless steel pan, overload protection, transportation lock, slip-resistant/adjustable feet, leveling bubble, stability indicator, low battery indicator, and auto shut-off. Applications include weighing, parts counting, percent weighing, and checkweighing.
All the balances come with an AC power adapter and can also operate on batteries (not included) allowing them to be transported and used where a power outlet is not available.
50,400.00 - ₹150,000.00INR / Each
21 variations of this product are available.
See All Product Options Below
21 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (g) Width (in)
Availability Pricing
Ohaus NV222 Navigator™ Portable Balance, 220 g x 0.01 g WW-11803-58 Mfr # NV222
COMPARE
220 0.01 8
₹86,100.00INR / Each
Ohaus NV422 Navigator™ Portable Balance, 420 g x 0.01 g WW-11803-59 Mfr # NV422
COMPARE
420 0.01 8
₹109,000.00INR / Each
Ohaus NV221 Navigator™ Portable Balance, 220 g x 0.1 g WW-11803-60 Mfr # NV221
COMPARE
220 0.1 8
₹50,400.00INR / Each
Ohaus NV621 Navigator™ Portable Balance, 620 g x 0.1 g WW-11803-61 Mfr # NV621
COMPARE
620 0.1 8
₹73,800.00INR / Each
Ohaus NV1201 Navigator™ Portable Balance, 1,200 g x 0.1 g WW-11803-62 Mfr # NV1201
COMPARE
1200 0.1 9 1/2
₹83,500.00INR / Each
Ohaus NV2201 Navigator™ Portable Balance, 7.5" x 5.7" Pan, 2,200 g x 0.1 g WW-11803-63 Mfr # NV2201
COMPARE
2200 0.1 9 1/2
₹92,700.00INR / Each
Ohaus NVT2201 Navigator™ Portable Balance, 9.1" x 6.9" Pan, 2,200 g x 0.1 g WW-11803-64 Mfr # NVT2201
COMPARE
2200 0.1 9 1/2
₹92,700.00INR / Each
Ohaus NVT4201 Navigator™ Portable Balance, 4,200 g x 0.1 g WW-11803-65 Mfr # NVT4201
COMPARE
4200 0.1 9 1/2
₹116,000.00INR / Each
Ohaus NVT2200 Navigator™ Portable Balance, 2,200 g x 1 g WW-11803-66 Mfr # NVT2200
COMPARE
2200 1 9 1/2
₹87,600.00INR / Each
Ohaus NVT6200 Navigator™ Portable Balance, 6,200 g x 1 g WW-11803-67 Mfr # NVT6200
COMPARE
6200 1 9 1/2
₹116,000.00INR / Each
Ohaus NVT12000 Navigator™ Portable Balance, 12,000 g x 1 g WW-11803-68 Mfr # NVT12000
COMPARE
12000 1 9 1/2
₹125,000.00INR / Each
Ohaus NVT22000 Navigator™ Portable Balance, 22,000 g x 1 g WW-11803-69 Mfr # NVT22000
COMPARE
22000 1 9 1/2
₹150,000.00INR / Each
Ohaus NV123  Navigator Portable Toploading Balance 120 g x 0.001 g  with Touchless Sensors WW-55000-18 Mfr # NV123
COMPARE
120 0.001 8
₹95,700.00INR / Each
Ohaus NV223  Navigator Portable Toploading Balance  220 g x 0.001 g with Touchless Sensors WW-55000-19 Mfr # NV223
COMPARE
220 0.001 8
₹107,000.00INR / Each
Ohaus NV323  Navigator Portable Toploading Balance 320 g x 0.001 g with Touchless Sensors WW-55000-20 Mfr # NV323
COMPARE
320 0.001 8
₹117,000.00INR / Each
Ohaus NV622 Navigator Portable Toploading Balance 620 g x 0.01 g with Touchless Sensors WW-55000-21 Mfr # NV622
COMPARE
620 0.01 8
₹112,000.00INR / Each
Ohaus NV1202  Navigator Portable Toploading Balance 1,200 g x 0.01 g with Touchless Sensors WW-55000-22 Mfr # NV1202
COMPARE
1200 0.01 8
₹124,000.00INR / Each
Ohaus NV2202  Navigator Portable Toploading Balance 2,200 g x 0.01 g with Touchless Sensors WW-55000-23 Mfr # NV2202
COMPARE
2200 0.01 8
₹131,000.00INR / Each
Ohaus NV3202  Navigator Portable Toploading Balance NV3202 AM 3,200 g x 0.01 g with Touchless Sensors WW-55000-24 Mfr # NV3202
COMPARE
3200 0.01 8
₹137,000.00INR / Each
Ohaus NVT6201 Navigator Portable Toploading Balance 6,200 g x 0.1 g with Touchless Sensors WW-55000-25 Mfr # NVT6201
COMPARE
6200 0.1 9 1/2
₹117,000.00INR / Each
Showing 1 - 20 of 21