This product is now discontinued, View Related Products
Ohaus® HH Handheld Balances
Convenient, compact and portable weighing in the palm of your hand
  • Backlit display for easy viewing in low light
  • Protective snap-on cover
Small enough to fit in a pocket, this electronic scale is great for prep work and field experiments.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Battery Capacity (g) Readability (g)
Availability Pricing
Ohaus HH120-D Standard Handheld Balance, 90g/120g WW-11013-50 Mfr # HH120D
COMPARE
120 0.2
Call For Price
Ohaus HH320-0W0 High-Performance Hh Handheld Balance, 320g Capacity WW-11013-51 Mfr # HH320
COMPARE
2 x AAA (included) 320 0.1
Call For Price
Ohaus HH120-0W0 High-Performance Hh Handheld Balance, 120g Capacity WW-11013-52 Mfr # HH120
COMPARE
2 x AAA (included) 120 0.1