This product is now discontinued, View Related Products
Oakton® Waterproof CON 610 Conductivity/TDS/Salinity/Resistivity Meters
Measure conductivity, TDS, salinity, resistivity and temperature in one easy-to-use meter!
  • Stores up to 500 data points with time-and-date stamp for GLP compliance
  • Programmable 'calibration due' date for increased data integrity
  • Wireless download of data via IrDA port
These top-of-the-line waterproof meters offer advanced measurement capabilities to meet all of you testing needs. The IP67-rated housing allows use in dirty and we environments without harm to the meter. Store up to 500 data points and download to computer or printer with the infrared (IrDA) wireless technology — no need for additional cords. User-selectable 'calibration due' and set-point alarms, automatic or manual standards recognition, and single or multiple point calibration provide flexibility in meeting you data quality objectives. The large backlit, dot matrix LCD shows the measured value and units, temperature, time, battery life, and menu options.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Waterproof Con/TDS/Res/Sal/Temp Meter w/ Probe
Call For Price
COMPARE
Waterproof Con/TDS/Res/Sal/Temp Meter w/ probe and NIST Cal
Call For Price
COMPARE
Waterproof Con/TDS/Res/Sal/Temp Meter Only
Call For Price
COMPARE
Waterproof Con/TDS/Res/Sal/Temp Meter w/ NIST Cal
Call For Price
COMPARE
Waterproof Con/TDS/Res/Sal/Temp Meter Kit
Call For Price
COMPARE
Waterproof Con/TDS/Res/Sal/Temp Meter Kit w/ NIST Cal