This product is now discontinued, View Related Products
Oakton® Traceable® pH Standard Buffers with Calibration, Individually-Tested
Individually-tested and serialized, 16 oz (473 mL), bottles
  • Get accurate, reliable measurements with included NIST-traceable calibration
  • Choose from clear or standard-colored buffers
Economical 473-mL bottles are individually-tested, serialized, and include a NIST-traceable calibration report. Choose from clear solutions, or color-standardized solutions: red is 4.005, yellow is 7.000, or blue is 10.012 pH. Buffers are supplied with NIST-traceable calibration report stating accuracy at 25°C to ±0.010 pH.
Certified Reference Materials (CRM) meet federal, state, and or local agencies’ strictest mandates, and deliver exact calibration results for any pH meter. A2LA ISO/IEC 17025:2005 (1750.01) (Calibration Laboratory) and ISO Guide 34 (Certified Reference Material Producer) provide highest achievable levels of product production, documentation, and accuracy. Additional accreditations include ISO 31 (content) and ISO 35 (statistical analysis). ISO 9001 ensures that world-class product standards for Materials are always met.
Traceable® products are calibrated to NIST-traceable standards and ready to use as soon as you receive it. Each product goes through an intense vetting process before it can carry the Traceable name, and it includes NIST-traceable certification. T
his means that a 17025-accredited calibration test lab has confirmed that the product meets or exceeds all stated specifications, saving you both time and money by not having to calibrate separately. NIST-traceable calibration also helps you meet quality standards and regulatory compliance for ISO, FDA, cGMP, VFC, CAP, CLIA and Joint Commission requirements.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Buffer Value Description
Availability Pricing
COMPARE
4.005 Traceable® pH Standard Buffer with Calibration, Clear, pH 4.005; 473 mL
Call For Price
COMPARE
7 Traceable® pH Standard Buffer with Calibration, Clear, pH 7.000; 473 mL
Call For Price
COMPARE
10.012 Traceable® pH Standard Buffer with Calibration, Clear pH 10.01; 473 mL
Call For Price
COMPARE
4.005 Traceable® pH Standard Buffer with Calibration, Red, pH 4.005; 473 mL
Call For Price
COMPARE
7 Traceable® pH Standard Buffer with Calibration, Yellow, pH 7.000; 473 mL
Call For Price
COMPARE
10.01 Traceable® pH Standard Buffer with Calibration, Blue, pH 10.01; 473 mL