This product is now discontinued, View Related Products
Oakton® Temp-10 Single-Input Thermocouple Thermometers
Heavy-duty thermometer offers versatility and accuracy in a compact design
  • 3-way hands-free operation.
  • Large easy-to-read screen with back light.
  • Rugged armor—built to withstand the toughest conditions.
  • Easy-to-use automatic field calibration ensures accurate readings.
  • Probes are sold separately
These robust and reliable meters work in any environment from the lab to the production floor. The sturdy armored construction protects against knocks and jars. All models are ergonomically designed for both large and small hands alike. The sealed keypad and ABS plastic case meet IP54 standards for splash resistance. Select from the three most popular thermocouple types—J, K, or T—to meet all your basic temperature measurement needs.
Features include a large, easy-to-read illuminated display; an easy-to-use five button control panel; Min/Max and Hold functions; °F/°C selection; auto shutoff; and low-battery indicator. Each thermometer is individually factory calibrated. Automatic field calibration is easily performed: simply place the probe in a container packed with ice, fill with water, then press the “CAL” button—meter
automatically recognizes the freezing point.
OAKTON’s exclusive “3-WAY HANDS-FREE” capability allows “hands-free” use—use the built-in stand on a table top, attach the meter to metal objects through magnets inserted into the armor boot, or hang from a pipe or belt using the hook-and-loop strap. Meters come standard with the integral stand;
the “hands-free” option adds magnets and strap (order separately).
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° C) Types Of Probes Accepted Description
Availability Pricing
COMPARE
1200 Type J Temp-10 Type J Thermocouple Thermometer with Boot
Call For Price
COMPARE
1200 Type J Temp-10 Thermocouple Meter; Type J, with NIST-Traceable Calibration
Call For Price
COMPARE
1372 Type K Temp-10 Type K Thermocouple Thermometer with Boot
Call For Price
COMPARE
1372 Type K Temp-10 Thermocouple Meter; Type K, with NIST-Traceable Calibration
Call For Price
COMPARE
1372 Type T Temp-10 Type T Thermocouple Thermometer with Boot
Call For Price
COMPARE
1372 Type T Temp-10 Thermocouple Meter; Type T, with NIST-Traceable Calibration