Oakton® Reference Fill Solutions
Effortless refilling
  • Choose your solution: 3.8 KCI with AgCI for Ag/AgCl electrodes or 4 KCI for calomel
  • Sold in packs of two 125-mL bottles
Choose from two types of reference fill solutions: 3.8 M KCl saturated with AgCl for AgCl/AgCl electrodes, or 4 M KCl for calomel (Hg/Hg22) electrodes. Both solutions come in a pack of two 125-mL — enough to fill approximately 25 electrodes.
5,300.00 - ₹13,800.00INR / Pkg of 2
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Buffer Value Volume (mL)
Availability Pricing
COMPARE
125
₹13,800.00INR / Pkg of 2
COMPARE
125
₹5,300.00INR / Pkg of 2