Oakton® pH Buffer Solution Packs
Save over 25% on pack compared to ordering separately
  • All Oakton pH buffers are traceable to NIST standard reference materials
  • Buffer solutions help you calibrate pH instruments to ensure accuracy
  • If unopened, shelf life is 2 years
• Oakton® pH Buffer Bottles have pH/temperature tables printed on the labels for accurate calibrations
• Manufactured under ISO 9000 quality guidelines and traceable to NIST standards
• If unopened, shelf life is 2 years; once opened, use pH 4 and pH 7 within 90 days, pH 10 within 30 days
• Store buffers away from direct sunlight.
4,300.00 - ₹44,700.00INR / Pkg of 3
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Buffer Kit Values Volume (mL) Description
Availability Pricing
COMPARE
1.68, 4.01, 7.00, 10.01, and 12.46 60 Buffer Pack; 60 mL of each pH 1.68, 4.01, 7.00, 10.01, and 12.46
₹17,100.00INR / Pkg of 5
COMPARE
4.01, 7.00, and 10.01 500 Buffer Pack; 500 mL of each pH 4.01, 7.00, and 10.01
₹4,300.00INR / Pkg of 3
COMPARE
4.01, 7.00, and 10.01 500 Buffer Pack; 10 x 500 mL of each pH 4.01, 7.00, and 10.01
₹44,700.00INR / Pkg of 30