Oakton® Ion Selective Electrodes
A wide variety of ion selective electrodes are availabe to suit a wide array of applications
  • Epoxy-body types are either single- or double-junction
  • Glass-body type is double junction
Select from four electrode types: membrane, solid-state, gas sensing, and glass bulb; typical response time is 20 to 30 seconds.
25,400.00 - ₹71,700.00INR / Each
22 variations of this product are available.
See All Product Options Below
22 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Electrode Body Material Electrode Type Ion-Selective Electrode
Availability Pricing
COMPARE
Epoxy Single junction, gas sensing Ammonia
₹62,100.00INR / Each
COMPARE
Epoxy Single junction, membrane Ammonium
₹60,100.00INR / Each
COMPARE
Glass Double junction, membrane Ammonium
₹71,700.00INR / Each
COMPARE
Epoxy Single junction, solid state Chloride
₹51,600.00INR / Each
COMPARE
Glass Double junction, solid state Chloride
₹60,700.00INR / Each
COMPARE
Epoxy Single junction, solid state Fluoride
₹60,700.00INR / Each
COMPARE
Glass Double junction, solid state Fluoride
₹60,700.00INR / Each
COMPARE
Epoxy Single junction, membrane Nitrate
₹71,700.00INR / Each
COMPARE
Glass Double junction, membrane Nitrate
₹71,700.00INR / Each
COMPARE
Epoxy Single junction, solid state Silver/Sulfide
₹51,100.00INR / Each
COMPARE
Glass Double junction, solid state Silver/Sulfide
₹60,900.00INR / Each
COMPARE
Epoxy Single junction Sodium
₹49,100.00INR / Each
COMPARE
Glass Double junction Sodium
₹69,900.00INR / Each
COMPARE
Epoxy Single junction, membrane Water hardness Ca/Mg
₹60,100.00INR / Each
COMPARE
Glass Double junction, membrane Water hardness Ca/Mg
₹71,300.00INR / Each
COMPARE
Epoxy Double junction, membrane Ammonium
₹44,300.00INR / Each
COMPARE
Epoxy Double junction, solid state Chloride
₹36,400.00INR / Each
COMPARE
Epoxy Double junction, solid state Fluoride
₹44,200.00INR / Each
COMPARE
Epoxy Double junction, membrane Nitrate
₹43,700.00INR / Each
COMPARE
Epoxy Double junction, solid state Silver/Sulfide
₹37,400.00INR / Each
Showing 1 - 20 of 22