Oakton® EcoTestr™ Conductivity Waterproof Pocket Testers
Designed to be durable and easy to use
  • View measurements clearly on large LCD
  • Rugged IP67-rated housing is waterproof, dustproof, and floats in water
  • Leakproof cap with fill line functions as a sample/calibration and hydrating cup
  • Secure—attach a lanyard to prevent losing or dropping in the field
  • Auto shutoff extends battery life
These economical, simple-to-use pocket testers quickly measure conductivity, TDS, or salinity levels accurately while simultaneously displaying the temperature. A large dual-line display makes viewing measurements easy and features indicators for measurement stability, calibration, and battery life.
Additional features include a long-lasting, durable electrode with polymer reference electrolyte, auto recognition of calibration standards, and self-diagnostics. The IP67-rated housing features an easy-to-grip design and floats if accidentally dropped in water when sensor cap is on. The testers, powered by four AAA batteries, can operate up to 400 hours continuously. The unit will auto power-off after 8 minutes of nonuse to conserve power.
8,900.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Calibration Temperature compensation Description
Availability Pricing
COMPARE
1 or 2 points Automatic Waterproof Conductivity Tester
₹8,900.00INR / Each
COMPARE
1 or 2 points Automatic Waterproof TDS Tester
₹8,900.00INR / Each
COMPARE
1 point Automatic Waterproof Salinity Tester
₹8,900.00INR / Each