Oakton® Conductivity Calibration Pouches
Sealed for freshness and easy travel
  • Tear off a corner, or the entire top—opens wide enough for all probe types
  • Notched corners for easy opening from either side
Single-use 20-mL pouches are sealed to ensure freshness—take these convenient standard solutions anywhere! Don't worry about transferring solution into another container—calibrate directly from the pouch.
A µS-to-ppm conversion table is printed on each pack for quick reference.
6,800.00 - ₹16,800.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Conductivity Value (µS) Description
Availability Pricing
COMPARE
84 Conductivity Calibration Pouches, 84 uS; Box of 20 Pouches
₹12,800.00INR / Pkg of 20
COMPARE
10 conductivity calibration pouches, 10 µS, box of 20 pouches
₹16,800.00INR / Each
COMPARE
447 conductivity calibration pouches, 447 µS, box of 20 pouches
₹6,800.00INR / Pkg of 20
COMPARE
1413 conductivity calibration pouches, 1413µS, box of 20 pouches
₹11,800.00INR / Pkg of 20
COMPARE
15000 conductivity calibration pouches, 15,000 µS, box of 20 pouches
₹7,400.00INR / Pkg of 20
COMPARE
12880 Conductivity Calibration Pouches, 12.88 mS; Box of 20 Pouches
₹11,800.00INR / Pkg of 20