Oakton® Conductivity and TDS Standards
Ideal for laboratory and field analysis
  • All Oakton Conductivity and TDS Standards are traceable to NIST standard reference materials
  • Solutions help you calibrate conductivity instruments to ensure accuracy
  • Standard values given at 25°C
KCl, NaCl and 442 conversions printed on the bottle for ease of use (excluded on 60-mL bottle, but included in insert). Temperature corrections are printed on the bottle, with certified values listed in tables and referenced to 25°C (excluded on 60-mL bottle, but included in insert). Manufactured under ISO 9000 quality guidelines and traceable to NIST standards. If unopened, shelf life is 1 year. If opened recommended shelf life is 4 months. Conductivity options range from 1 µS/cm to 200,000 µS/cm.
1,400.00 - ₹12,000.00INR / Pint of 1
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Conductivity Value (µS) Volume (mL)
Availability Pricing
COMPARE
12880 500
₹4,000.00INR / Pint of 1
COMPARE
1500 500
₹1,400.00INR / Each
COMPARE
84 500
₹3,000.00INR / Pint of 1
COMPARE
1413 500
₹4,000.00INR / Pint of 1
COMPARE
2764 500
₹2,000.00INR / Pint of 1
COMPARE
23 500
₹5,000.00INR / Each
COMPARE
2070 500
₹2,000.00INR / Each
COMPARE
80000 500
₹3,900.00INR / Each
COMPARE
100 60
₹6,400.00INR / Pkg of 5
COMPARE
1413 60
₹12,000.00INR / Pkg of 5
COMPARE
12880 60
₹6,400.00INR / Pkg of 5
COMPARE
111900 60
₹4,400.00INR / Pkg of 5
COMPARE
447 500
₹2,000.00INR / Each
COMPARE
15000 500
₹2,000.00INR / Each
COMPARE
8974 500
₹2,000.00INR / Each
COMPARE
10 500
Call For Price
COMPARE
5 500
Call For Price
Showing 1- 17 of 17