Oakton® Combination pH Electrodes
Outstanding value at an economical cost!
  • Compatible with all Oakton brand pH meters
Oakton combination electrodes are available in a variety of styles to meet the needs of your application. Epoxy electrodes are impact resistant and provide excellent durability for field applications. Glass electrodes withstand aggressive chemicals. Choose junction and tip style based on the needs of your application. All electrodes feature Ag/AgCl reference cells.
7,600.00 - ₹35,600.00INR / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Applications Electrode Type Description
Availability Pricing
COMPARE
General-purpose measurements Single junction General-Purpose pH Probe, Sealed/SJ/Epoxy/Direct Connect; BNC
₹9,100.00INR / Each
COMPARE
Dirty Samples Double junction General-Purpose pH Probe, Sealed/DJ/Epoxy/Direct Connect; BNC
₹10,500.00INR / Each
COMPARE
Food-semi solids Single junction pH Probe, Sealed/SJ/Glass/Spear-Tip/3 ft Cable; BNC
₹18,000.00INR / Each
COMPARE
Dirty Samples Double junction General-Purpose pH Probe, Sealed/DJ/Epoxy/3 ft Cable; BNC
₹10,400.00INR / Each
COMPARE
General-purpose measurements Double junction General-Purpose pH Probe, Refillable/DJ/Glass/3 ft Cable; BNC
₹18,700.00INR / Each
COMPARE
High pH or sodium Single junction pH Probe, Sealed/SJ/Epoxy/High pH Sodium/3 ft Cable; BNC
₹7,600.00INR / Each
COMPARE
High pH or sodium Double junction pH Probe, Sealed/DJ/Epoxy/High pH Sodium/3 ft Cable; BNC
₹11,700.00INR / Each
COMPARE
Dirty Samples-flushable Double junction pH Probe, Refillable/Flushable PTFE DJ/Epoxy; BNC
₹32,600.00INR / Each
COMPARE
Dirty Samples Double junction General-Purpose ORP Probe, Sealed/DJ/Epoxy/3 ft Cable; BNC
₹20,900.00INR / Each
COMPARE
Food-meats Double junction flat-surface, glass, double junction,BNC, pH electrode
₹35,600.00INR / Each
COMPARE
Food-meats Double junction flat-surface, epoxy, gel,BNC, pH electrode
₹28,900.00INR / Each
Showing 1- 11 of 11