Noshok Platinum RTD Temperature Switch and Transmitter

FEATURES
  • Wide variety of temperature ranges and connections
  • CE compliant
  • Digital calibration
  • Proven Platinum 100 ohm sensor

APPLICATIONS
  • Industrial machinery
  • Storage tanks
  • Water systems
  • HVAC systems

The Series 800 temperature transmitter provides a previously unequalled leel of performance, reliability and simplicity in one package. Utilizing proven 100 ohm platinum resistance technology coupled with a digital ASIC electronics insures reliability and maximum performance. A rigorous test and inspection process is performed prior to shipment to further insure 100% "out of the box" reliability.
86,500.00 - ₹88,800.00INR / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Loop Power Supply
Availability Pricing
NOSHOK 800-0/200-1-1-8-8-040-6 RTD Temperature Transmitter, 0/200F, +/-0.5% Fs Accuracy, 4" Stem WW-94713-76 Mfr # 800-0/200-1-1-8-8-040-6
COMPARE
₹86,500.00INR / Each
NOSHOK 800-0/200-1-1-8-8-060-6 RTD Temperature Transmitter; 0 to 200F, 6" Stem WW-94713-77 Mfr # 800-0/200-1-1-8-8-060-6
COMPARE
₹88,800.00INR / Each
NOSHOK 800-40/120-1-1-8-8-040-6 RTD Temperature Transmitter; -40 to 120F, 4" Stem WW-94713-80 Mfr # 800-40/120-1-1-8-8-040-6
COMPARE
₹86,500.00INR / Each
NOSHOK 800-50/500-1-1-8-8-040-6 RTD Temperature Transmitter; 50 to 500F, 4" Stem WW-94713-84 Mfr # 800-50/400-1-1-8-8-040-6
COMPARE
₹86,500.00INR / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat