This product is now discontinued, View Related Products
Noshok 400 Series Precision Needle Valves
Hard-seat design
  • Metal-to-metal hard seat design is 100% helium leak-tested to 1 x 10–4 mL/s at 200 psi
  • 10,000 psi pressure rating at 200°F (93°C)
  • Blowout-proof stem
Series 400 needle valves are high quality instrument grade valves. They are the right choice when your application calls for positive shut-off or precise flow control. They work effectively with most gases and liquids.
Call For Price
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Cv Inlet Port Outlet Port
Availability Pricing
NOSHOK 404-FFAC NeedleValve,HardSeat Steel Female-Female 1/2" NPT Female-Female Angle WW-68430-19 Mfr # 404-FFAC
COMPARE
0.44 1/2\" NPT(F) 1/2\" NPT(F)