North Sciences PR Series Lab/Pharmacy Refrigerators
Biopharmaceutical refrigerators with precision temperature control and cloud-based monitoring for total sample security
  • Features include leveling feet, interior shelves, and rear stoppers Forced-air circulation with special air ducts guarantees inner temperature consistency Unique evaporator design results in better uniformity with no ice build-up
The PR Series high-performance, biopharmaceutical refrigerators offer superior temperature uniformity and outstanding reliability. They are designed to store pharmaceuticals, medicine, vaccines, biological materials, testing reagents, and laboratory materials. The controller allows the inner temperature to be adjusted by increments of 0.1° with multiple sensors to display and accurately control the temperature. A safety door lock prevents unauthorized access. Auto defrost function offers reliable performance in high ambient temperature and high humidity areas.
612,000.00 - ₹638,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature - Refrigerator (° F) Min Temperature - Refrigerator (° F) Power (VAC)
Availability Pricing
North Sciences PR Series Lab/Pharmacy Refrigerator, 2°C to 8°C, 395 L, 110 V WW-44260-98 Mfr # PR-0400-NA
COMPARE
46.5 36 120
₹638,000.00INR / Each
North Sciences PR Series Lab/Pharmacy Refrigerator, 2°C to 8°C, 395 L, 220 V WW-44260-99 Mfr # PR-0400-EU
COMPARE
46.5 36 220
₹612,000.00INR / Each