North Sciences Messenger™ Portable Ultra-Low Temperature Freezer
True portable ULT freezer solutions for delivering vaccines at ultra-low temperature without risks
  • Ideal for clinical trials and biologic drug delivery
  • PT100 thermo-sensor ensures temperature control accuracy of the whole box and avoids temperature fluctuation
  • Digital locks protect samples
  • Integrated high pressure foam, VIP (vacuum insulation panel), double layer submersible doors, and double layer sealing rings ensure ultra-low thermal leakage
  • Advanced Stirling technology offers fast cooling, no fear of bumps and shakes, and maintenance free life up to 50,000 hours
The ultra-low temperature portable freezers are specially designed for critical sample shipments. It is small, light in weight, and reliable. They have a built-in, large capacity lithium battery which can be used to transport the samples for more than 1 hour without any external power source. The portable freezers use 100% natural refrigerants and piston-based technology with no compressors or moving parts.
1,596,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Freezer Capacity (cu ft) Min Temperature - Freezer (° C)
Availability Pricing
North Sciences Messenger™ 1 Portable ULT Freezer, -80°C to 30°C, 1 L, 12-24 V WW-44260-96 Mfr # DM8501
COMPARE
0.035 -86
₹1,596,000.00INR / Each