This product is now discontinued, View Related Products
North by Honeywell North Adaptec™ Safety Glasses
Customized adjustability fits all faces
  • Smoke tint lens minimizes outdoor sunlight and glare HydroShield long-life antifog coating helps keep lenses clear Multi-adjustable frame design for a comfortable and snug fit
With three different lens sizes to choose from, these scratch resistant polycarbonate vented glasses are sure to give you a more customized fit. They feature an adjustable lens angle and temple tension, along with a nose bridge, serving as added safeguards.
Compliance: Certified to meet ANSI+ CSA Z94.3 (standard).
21,100.00 - ₹23,000.00INR / Box of 10
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Frame Color Lens Coating Lens Type
Availability Pricing
Uvex by Honeywell T5900WTKHS North Adaptec Safety Glasses, Translucent Black Frame, Smoke, HydroShield Antifog, Wide Lens; 10/BX WW-86498-03 Mfr # T5900WTKHS
COMPARE
Translucent Black Antifog Wide
₹23,000.00INR / Box of 10
Uvex by Honeywell T5900WBLHS North Adaptec Safety Glasses, Dark Blue Frame, Smoke, HydroShield Antifog, Wide Lens; 10/BX WW-86498-04 Mfr # T5900WBLHS
COMPARE
Dark Blue Antifog Wide
Call For Price
Uvex by Honeywell T5900NTKHS North Adaptec Safety Glasses,Translucent  Black Frame, Smoke, HydroShield Antifog, Narrow Lens; 10/BX WW-86498-05 Mfr # T5900NTKHS
COMPARE
Translucent Black Antifog Narrow
₹21,100.00INR / Box of 10
Uvex by Honeywell T5900NBLHS North Adaptec Safety Glasses, Dark Blue Frame, Smoke, HydroShield Antifog, Narrow Lens; 10/BX WW-86498-06 Mfr # T5900NBLHS
COMPARE
Dark Blue Antifog Narrow
Call For Price
Uvex by Honeywell T5900LTKHS North Adaptec Safety Glasses, Translucent Black Frame, Smoke, HydroShield Antifog, Regular Lens; 10/BX WW-86498-07 Mfr # T5900LTKHS
COMPARE
Translucent Black Antifog Regular
₹23,000.00INR / Box of 10
Uvex by Honeywell T5900LBLHS North Adaptec Safety Glasses, Dark Blue Frame, Smoke, HydroShield Antifog, Regular Lens; 10/BX WW-86498-08 Mfr # T5900LBLHS
COMPARE
Dark Blue Antifog Regular
₹23,000.00INR / Box of 10