This product is now discontinued, View Related Products

Nor-Lake General Purpose Laboratory Refrigerators

Designed for the demanding laboratory
797,000.00 - ₹808,000.00INR / Each

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Refrigerator Capacity (cu ft)
Availability Pricing
Nor-Lake NSLR241WMW/0M Refrigerator, Manual Defrost 24 cu ft with Casters, 115V WW-44173-04 Mfr # NSLR241WMW/0M
COMPARE
115 24
Call For Price
Nor-Lake NSLR241WMW/5M Refrigerator, Manual Defrost 24 cu ft with Casters, 230V WW-44173-06 Mfr # NSLR241WMW/5M
COMPARE
230 24
Call For Price
Nor-Lake NSLR041WMW/0 Refrigerator, Automatic Defrost Undercounter 4.2 cu ft with Legs, 115V/60Hz WW-44173-56 Mfr # NSLR041WMW/0
COMPARE
115
Call For Price
Nor-Lake NSLR041WMW/0M Refrigerator, Manual Defrost Undercounter 4.2 cu ft with Legs, 115V/60Hz WW-44173-58 Mfr # NSLR041WMW/0M
COMPARE
Call For Price
Nor-Lake NSLR051WMW/0 Refrigerator, Automatic Defrost Undercounter 5.1 cu ft with Legs, 115V/60Hz WW-44173-60 Mfr # NSLR051WMW/0
COMPARE
230
₹808,000.00INR / Each
Nor-Lake NSLR051WMW/0M Refrigerator, Manual Defrost Undercounter 5.1 cu ft with Legs, 115V/60Hz WW-44173-62 Mfr # NSLR051WMW/0M
COMPARE
115
₹797,000.00INR / Each
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat