Nichrome V Non-Calibrated Inoculating Loops without Handles
Select, inoculate, or transfer microbiological samples
  • Nichrome wire composition cools quickly
  • Constructed of 26-gauge wire
Ideal to streak various liquids, suspensions, bacteria, and other specimens on to culture media and/or petri dishes for purposes of study and growth. Non-calibrated loops do not include a handle, order handle 14203-34 separately.
3,900.00 - ₹7,000.00INR / Pkg of 12
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
B and S Wire Gauge Loop Inside Diameter (mm) Loop Length (in)
Availability Pricing
Nichrome V Inoculating Loop without Handle, 26 Gauge, 2 mm ID; 12/Pk WW-14203-27 Mfr # 3512
COMPARE
26 2 3
₹3,900.00INR / Pkg of 12
Nichrome V Inoculating Loop without Handle, 26 Gauge, 3 mm ID; 12/Pk WW-14203-28 Mfr # 3513
COMPARE
26 3 3
₹4,500.00INR / Pkg of 12
Nichrome V Inoculating Loop without Handle, 26 Gauge, 4 mm ID; 12/Pk WW-14203-29 Mfr # 3514
COMPARE
26 4 3
₹7,000.00INR / Pkg of 12
Nichrome V Inoculating Loop without Handle, 26 Gauge, 5 mm ID; 12/Pk WW-14203-30 Mfr # 3515
COMPARE
26 5 3
₹4,500.00INR / Pkg of 12