Nichrome V Non-Calibrated Inoculating Loops and Needles with Handles
Select, inoculate, or transfer microbiological samples
  • Nichrome wire composition cools quickly
  • Constructed of Nichrome wire and aluminum handle
Ideal to streak various liquids, suspensions, bacteria, and other specimens on to culture media and/or petri dishes for purposes of study and growth. These loops and needles are 2-3/4” long plus the length of the handle. Choose from 24 gauge wide which is the most durable, 25 gauge wire for general use, or 26 gauge wire which is designed for rapid cooling.
8,600.00 - ₹15,100.00INR / Pkg of 12
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
B and S Wire Gauge Handle length (mm) Handle Type
Availability Pricing
Nichrome V Inoculating Loop with Handle, 6" L, Non-Insulated, 3 mm ID, 24 Gauge; 12/Pk WW-14203-20 Mfr # 3103
COMPARE
24 152 Non-Insulated; Loop
₹11,200.00INR / Pkg of 12
Nichrome V Inoculating Loop with Handle, 8" L, Non-Insulated, 4 mm ID, 24 Gauge; 12/Pk WW-14203-21 Mfr # 3124
COMPARE
24 203 Non-Insulated; Loop
₹10,000.00INR / Pkg of 12
Nichrome V Inoculating Loop with Handle, 8" L, Non-Insulated, 3 mm ID, 25 Gauge; 12/Pk WW-14203-22 Mfr # 3128
COMPARE
25 203 Non-Insulated; Loop
₹11,800.00INR / Pkg of 12
Nichrome V Inoculating Loop with Handle, 8" L, Insulated, 3 mm ID, 26 Gauge; 6/Pk WW-14203-23 Mfr # 3153
COMPARE
26 203 Insulated: Loop
₹8,600.00INR / Pkg of 6
Nichrome V Inoculating Loop with Handle, 8" L, Insulated, 2 mm ID, 26 Gauge; 6/Pk WW-14203-24 Mfr # 3152
COMPARE
26 203 Insulated: Loop
₹9,100.00INR / Pkg of 6
Nichrome V Inoculating Needle with Handle, 6" L, Non-Insulated, 24 Gauge; 12/Pk WW-14203-32 Mfr # 3000
COMPARE
24 152 Non-Insulated; Needle
₹15,100.00INR / Pkg of 12
Nichrome V Inoculating Needle with Handle, 8" L, Insulated, 24 Gauge; 6/Pk WW-14203-33 Mfr # 3060
COMPARE
24 203 Insulated; Needle
₹9,200.00INR / Pkg of 6
Showing 1- 7 of 7