This product is now discontinued, View Related Products
Neptune Low-Profile Right-Angle IBC Tote Mixers
Save the time and trouble of draining fluids from their containers—excellent for short- or long-term storage
  • Low-profile gear drive mixers fit between stacked semi-bulk shipping containers
  • Folding propeller fits through 2" (51 mm) opening—expands to 9" (229 mm) operating diameter
These gear-drive mixers are perfect for mixing the contents of semi-bulk shipping containers. Mount mixers directly to a 2" (51 mm) NPT coupling, or order the optional cap mounting flange to use mixer on any plastic screw cap on bulk mixing containers. Simply mount the flange to your bulk cap by drilling four holes, mixer attaches to the flange and you are ready to mix. Power requirement for TEFC motors is 115 to 230 VAC, 60 Hz, single phase.
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Max Torque (in-oz) Motor hp Motor Type
Availability Pricing
Neptune Bulk container mixer; 1/2 hp, 2" NPT(M) mount, 350 rpm, 115/230 VAC WW-50316-02 Mfr # RAB-3.0
COMPARE
384 1/2 TEFC
Call For Price
Neptune Low-Profile Right-Angle IBC Tote Mixer, 350 rpm, 1/2 HP; Air WW-50316-04 Mfr # RGT-1.4
COMPARE
384 1/2 Air-driven
Call For Price
Neptune Bulk container mixer; 1/2 hp, 2" NPT(M) mount, 350 rpm, air powered WW-50316-06 Mfr # RAB-4.0
COMPARE
384 1/2 Air-driven
Call For Price
Neptune Bulk container mixer; 3/4 hp, 2" NPT(M) mount, 350 rpm, 115/230 VAC WW-50316-10 Mfr # RAB-5.0
COMPARE
576 3/4 TEFC
Call For Price
Neptune Bulk container mixer; 1 hp, 2" NPT(M) mount, 350 rpm, 115/230 VAC WW-50316-14 Mfr # RAB-6.0
COMPARE
768 1 TEFC