This product is now discontinued, View Related Products
Neptune Handlebar Pail and Drum Air Mixers
Air control valve allows easy speed adjustment
Popular for mixing open and closed containers, these air mixers are suitable for mixing up to 55 gallons of thin or heavy liquids. Suitable for mixing mortar, drywall compounds, and similar materials.
Order trigger for start/stop control as item 50500-50 and air set 50500-32 separately.
238,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Impeller Size (in) Max Speed (rpm) Shaft Length (in)
Availability Pricing
Neptune Handlebar mixer; 5/8" dia x 30"L shaft, 4" dia folding two-blade propeller WW-50500-33 Mfr # HAM-3.0
COMPARE
4 2000 30
Call For Price
Neptune Handlebar mixer; 1/2" dia x 20.5"L shaft, 5" dia caged proller WW-50500-35 Mfr # HAJ-3.0
COMPARE
5 2000 20.5
Call For Price
Neptune Handlebar mixer; 1/2" dia x 40"L shaft, 5" dia caged proller WW-50500-37 Mfr # HAJ-3.1
COMPARE
5 2000 40
₹238,000.00INR / Each
HAF-3.0 Handlebar mixer; 5/8" dia x 21"L shaft, 3-1/4" dia two-blade folding propeller WW-50500-39 Mfr # HAF-3.0
COMPARE
2000 21
Call For Price
HAF-3.1 Handlebar mixer; 5/8" dia x 40"L shaft, 3-1/4" dia two-blade folding propeller WW-50500-41 Mfr # HAF-3.1
COMPARE
2000 40
Call For Price
HAF-3.2 Handlebar mixer; 5/8" dia x 40"L shaft, 8" dia six-blade folding propeller WW-50500-43 Mfr # HAF-3.2
COMPARE
8 2000 40