Sterile Disposable Reagent Reservoir
“V” shaped well design allows maximum reagent recovery
  • Easy to read graduation markings
  • Drip free pour-off spouts at every corner for spill free pouring
Sterile reagent reservoirs are constructed of white polystyrene, providing easy to read graduation markings located on the top edge with a fill line on inner walls to indicate volume levels. The V-shaped bottom allows for efficient and nearly complete sample withdrawal.
25,700.00INR / Pkg of 100
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Graduation Interval Max Volume (mL) Description
Availability Pricing
COMPARE
25- and 50-mL 55 Disposable Reagent Reservoir, Polystyrene, Sterile, 55 mL; 50/Pk
₹25,700.00INR / Pkg of 100