This product is now discontinued, View Related Products
Multichamber Sampling Lances for Powders and Granulate
Repeatable zone sampling made easy
  • Three or five chambers sample zones at a fixed depth
  • Removable tip simplifies cleaning
  • Gap between inner and outer tubes minimized to ensure smooth operation
  • Choose between aluminum, all stainless steel, or stainless steel and PTFE
These multichambered zone samplers feature sturdy metal construction with a handle that makes closing the chambers easy. Select strong and economical aluminum for noncorrosive applications, durable stainless steel for high purity and greater chemical resistance, or a combination of stainless steel and PTFE for a lighter-weight alternative to the all-stainless version.
Call For Price
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Length (in) Volume (mL)
Availability Pricing
Burkle 5316-0055 Multi layer sampling lance, SS/PTFE, 3 individual chambers, 55 cm; 14 mL per chamber WW-06041-34 Mfr # 5316-0055
COMPARE
21.625 14
Call For Price
Burkle 5316-0085 Multi layer sampling lance, SS/PTFE, 3 individual chambers, 85 cm; 17 mL per chamber WW-06041-35 Mfr # 5316-0085
COMPARE
33.5 17
Call For Price
Burkle 5316-0150 Multi layer sampling lance, SS/PTFE, 5 individual chambers, 150 cm; 17 mL per chamber WW-06041-36 Mfr # 5316-0150
COMPARE
59 17
Call For Price
Burkle 5316-3055 Multi layer sampling lance, SS, 3 individual chambers, 55 cm; 14 mL per chamber WW-06041-43 Mfr # 5316-3055
COMPARE
21.625 14
Call For Price
Burkle 5316-3085 Multi layer sampling lance, SS, 3 individual chambers, 85 cm; 17 mL per chamber WW-06041-44 Mfr # 5316-3085
COMPARE
33.5 17
Call For Price
Burkle 5316-3150 Multi layer sampling lance, SS, 5 individual chambers, 150 cm; 17 mL per chamber WW-06041-45 Mfr # 5316-3150
COMPARE
59 17
Call For Price
Burkle 5317-1058 Multi layer sampling lance, aluminim, 3 individual chambers, 55 cm; 14 mL per chamber WW-06041-48 Mfr # 5317-1058
COMPARE
21.625 14
Call For Price
Burkle 5317-1085 Multi layer sampling lance, aluminim, 3 individual chambers, 85 cm; 17 mL per chamber WW-06041-49 Mfr # 5317-1085
COMPARE
33.5 17
Call For Price
Burkle 5317-1150 Multi layer sampling lance, aluminim, 5 individual chambers, 150 cm; 17 mL per chamber WW-06041-50 Mfr # 5317-1150
COMPARE
59 17
Call For Price
Showing 1- 9 of 9