Multi Vortex-Genie Shaker
Provides true vortexing in almost any container up to 250 ml
  • Use for gentle, orbital type mixing or more vigorous vortexing
  • Durable and heavy zinc die casting with specially designed feet prevents ''walking''
These shakers hold multiple test tubes, vials, cylinders of various sizes, or up to six 250 ml vessel(racks included). Digital control and display of speed and time for reproducibility and accuracy, with true speed display. May be used in cold rooms and incubators.
444,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
Scientific Industries Genie SI-M236 Multi Shaker, 250 to 1000 rpm, 120 VAC WW-51402-30 Mfr # SI-M236
COMPARE
120
₹444,000.00INR / Each
Scientific Industries Genie SI-M246 Multi Shaker, 250 to 1000 rpm, 240 VAC WW-51402-32 Mfr # SI-M246
COMPARE
240
₹444,000.00INR / Each