Cole-Parmer® Multi-Element Probes for 500- and 750-Watt Ultrasonic Processors
Maximize productivity by processing numerous samples simultaneously
  • Probes process from 250 μL to 10 mL sample volume
  • Energy delivered to each tip is uniform with 2% variation
The multi-element probes screw into the converter in place of the 1⁄2" (13 mm) probe. The energy delivered to each tip is uniform with 2% variation. The spacing between the tips is 2 2⁄32" (18 mm). Coupler and stepped microtips measure 10 1⁄4" (260 mm) long. Probes process from 250 μL to 10 mL sample volume and consist of aluminum coupling and 4-, 8-, or 24-element probes.
216,000.00 - ₹271,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Sample Size (mL) Description
Availability Pricing
COMPARE
10 Ultrasonic Four-Element Probe; 1/8\" Dia
₹216,000.00INR / Each
COMPARE
10 Ultrasonic Eight-Element Probe; 1/8\" Dia
₹271,000.00INR / Each