This product is now discontinued, View Related Products
Multi-Element Imersion Heaters
Multi-Element Heaters Encased in 304 Stainless Steel or PTFE include a grounded solid-state heating element. Heaters are self-limiting for safer operation. PTFE-coated heaters are ideal for use in corrosive environments. The numbers of heating rods coming down from the polypropylene terminal head are dependent on the wattage of the heater: 300 Watt = 2 rods, 500 Watt = 3 rods, and 1000 Watt = 5 rods. Heaters include 3 ft wire and flexible conduit.
Call For Price
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Length (in) Max Temperature (° F) Power (VAC)
Availability Pricing
Process Technology VS.3111 Immersion Heater, Two 304 SS Rod s, 6" L Heated Area, 300 Watts, 120 V WW-36135-10 Mfr # VS.3111
COMPARE
450 120
Call For Price
Process Technology VS.3211 Immersion Heater, Two 304 SS Rod s, 6" L Heated Area, 300 Watts, 240 V WW-36135-15 Mfr # VS.3211
COMPARE
450 240
Call For Price
Process Technology VS.5111 Immersion Heater, Three 304 SS Rod s, 6" L Heated Area, 500 Watts, 120 V WW-36135-20 Mfr # VS.5111
COMPARE
450 120
Call For Price
Process Technology VS.5211 Immersion Heater, Three 304 SS Rod s, 6" L Heated Area, 500 Watts, 240 V WW-36135-25 Mfr # VS.5211
COMPARE
450 240
Call For Price
Process Technology VS.1111 Immersion Heater, Five 304 SS Rod s, 6" L Heated Area, 1000 Watts, 120 V WW-36135-30 Mfr # VS1111
COMPARE
450 120
Call For Price
Process Technology VS.1211 Immersion Heater, Five 304 SS Rod s, 6" L Heated Area, 1000 Watts, 240 V WW-36135-35 Mfr # VS1211
COMPARE
450 240
Call For Price
Process Technology VX.3111 Immersion Heater, Two PTFE Coated Rod s, 6" L Heated Area, 300 Watts, 120 V WW-36136-10 Mfr # VX.3111
COMPARE
450 120
Call For Price
Process Technology VX.3211 Immersion Heater, Two PTFE Coated Rod s, 6" L Heated Area, 300 Watts, 240 V WW-36136-15 Mfr # VX.3211
COMPARE
450 240
Call For Price
Process Technology VX.5111 Immersion Heater, Three PTFE Coated Rod s, 6" L Heated Area, 500 Watts, 120 V WW-36136-20 Mfr # VX.5111
COMPARE
450 120
Call For Price
Process Technology VX.5211 Immersion Heater, Three PTFE Coated Rod s, 6" L Heated Area, 500 Watts, 240 V WW-36136-25 Mfr # VX.5211
COMPARE
450 240
Call For Price
Process Technology VX1111 Immersion Heater, Five PTFE Coated Rod s, 6" L Heated Area, 1000 Watts, 120 V WW-36136-30 Mfr # VX1111
COMPARE
450 120
Call For Price
Process Technology VX1211 Immersion Heater, Five PTFE Coated Rod s, 6" L Heated Area, 1000 Watts, 240 V WW-36136-35 Mfr # VX1211
COMPARE
450 240
Call For Price
Showing 1- 12 of 12