This product is now discontinued, View Related Products
Mucksucker™ High-Suction Samplers
No leachable fillers or plasticizers
  • No glass parts to break for added safety and protection Easily identify layers through translucent material Easy to clean
High-suction syringe action is ideal for viscous fluids. Efficiently draws samples that are close to the bottom of the drum or tank. Comes with both a push/seal bottom and a coring bottom. Coring bottom features a wide mouth for handling sludge samples.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Mucksucker D8653259 drum and tank sampler, 3.5' long WW-99200-20 Mfr # D8653259
COMPARE
drum and tank sampler, 3.5' long
Call For Price
Mucksucker D8653260 Sampler, PTFE/FEP, 7 ft depth WW-99200-22 Mfr # D8653260
COMPARE
Sampler, PTFE/FEP, 7 ft depth