This product is now discontinued, View Related Products
Mounted In-Line Static Mixers
Mounted mixers can be used with any compression fitting with the same tubing OD.
10,800.00 - ₹19,300.00INR / Pkg of 10
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Mixer Length (in) Mounting Type No. Of Elements
Availability Pricing
Koflo 5/16-12PPT Mounted in-line mixer; 5/16" tube OD, 12 elements, 10/pack WW-04668-02 Mfr # 5/16-12PPT
COMPARE
3.5 0.3125\" plain end 12
₹10,800.00INR / Pkg of 10
Koflo 3/8-12PPT Mounted in-line mixer; 3/8" tube OD, 12 elements, 10/pack WW-04668-04 Mfr # 3/8-12PPT
COMPARE
4.25 0.375\" plain end 12
₹11,200.00INR / Pkg of 10
Koflo 1/2-12PPT Mounted in-line mixer; 1/2" tube OD, 12 elements, 10/pack WW-04668-06 Mfr # 1/2-12PPT
COMPARE
6.5 0.5\" plain end 12
₹12,500.00INR / Pkg of 10
Koflo 5/8-12PPT Mounted in-line mixer; 5/8" tube OD, 12 elements, 10/pack WW-04668-08 Mfr # 5/8-12PPT
COMPARE
8.5 0.625\" plain end 12
₹17,100.00INR / Pkg of 10
Koflo 5/16-24PPT Mounted in-line mixer; 5/16" tube OD, 24 elements, 10/pack WW-04668-12 Mfr # 5/16-24PPT
COMPARE
5.75 0.3125\" plain end 24
₹11,700.00INR / Pkg of 10
Koflo 3/8-24PPT Mounted in-line mixer; 3/8" tube OD, 24 elements, 10/pack WW-04668-14 Mfr # 3/8-24PPT
COMPARE
7.25 0.375\" plain end 24
₹12,300.00INR / Pkg of 10
Koflo 1/2-24PPT Mounted in-line mixer; 1/2" tube OD, 24 elements, 10/pack WW-04668-16 Mfr # 1/2-24PPT
COMPARE
12.625 0.5\" plain end 24
₹15,000.00INR / Pkg of 10
Koflo 5/8-24PPT Mounted in-line mixer; 5/8" tube OD, 24 elements, 10/pack WW-04668-18 Mfr # 5/8-24PPT
COMPARE
14.25 0.625\" plain end 24
₹19,300.00INR / Pkg of 10
Showing 1- 8 of 8