This product is now discontinued, View Related Products
Motic Moticam Digital Microscope Cameras with Software
Fast plug-and-play cameras relay images from your microscope to your computer or monitor
  • Supplied software measures specimens and logs each image with automatic reference number, date, and time
Capture electronic images and videos by converting your compound or stereo microscope into a powerful digital imaging system. These affordable digital cameras capture still or video images and instantly transmit them for real-time full screen viewing. Timed-interval shots provide time lapse observation—save, manipulate, print, and send!
Rapid view algorithms let you display at a faster frame rate while still being able to capture images at the highest resolution. These cameras are also great for capturing images of objects too large for a microscope stage. The included macro viewing tube is a stand that sits on the bench and holds the camera above a specimen.
Call For Price
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Image Sensor Output Resolution
Availability Pricing
Motic Instruments Moticam 1 Digital Microscope Camera, 1/4" CMOS WW-03906-11 Mfr # 1100600100601
COMPARE
1/4\" CMOS USB 0.48 MP
Call For Price
Motic Instruments Moticam 1SP Digital Microscope Camera, 1/3" CMOS WW-03906-12 Mfr # 1100600100641
COMPARE
1/3\" CMOS USB 1.3 MP
Call For Price
Motic Instruments Moticam 2 Digital Microscope Camera, 1/3" CMOS WW-03906-13 Mfr # 1100600100611
COMPARE
1/3\" CMOS USB 2.0 MP
Call For Price
Motic Instruments Moticam 3 Digital Microscope Camera, 1/2" CMOS WW-03906-14 Mfr # 1100600100791
COMPARE
1/2\" CMOS USB 3.0 MP
Call For Price
Motic Instruments Moticam 5 Digital Microscope Camera, 1/2.5" CMOS WW-03906-15 Mfr # 1100600100631
COMPARE
1/2.5\" CMOS USB 5 MP
Call For Price
Motic Instruments Moticam 10 Digital Microscope Camera, 1/2.5" CMOS WW-03906-16 Mfr # 1100600101211
COMPARE
1/2.5\" CMOS USB 10.0 MP
Call For Price
Motic Instruments Moticam 580 Digital Microscope Camera, 1/2.5" CMOS WW-03906-17 Mfr # 1100600100651
COMPARE
1/2.5\" CMOS USB, HDMI, SD card, analog video 5 MP
Call For Price
Motic Instruments Moticam X Digital Microscope Camera, Wi-Fi, 1/3" CMOS WW-03906-18 Mfr # 1100600100691
COMPARE
1/3\" CMOS Wi-Fi 2.0 MP
Call For Price
Showing 1- 8 of 8