This product is now discontinued, View Related Products
Motic Inverted Microscopes
Color-corrected infinity optical system maximizes clarity and scale
The working distance of these inverted microscopes is ample for larger culture vessels with or without the 0.30 N.A. condenser attached. The precentered system features a removable condenser allowing for flasks and Petri dishes as high as 184 mm to be accommodated on the stage.
The included CCIS phase Plan achromatic objectives coupled with the phase slider provides outstanding contrast images for even the most demanding applications. Semi-circular hole in the glass and metal stage inserts makes changing objectives easy without having to take the specimen off the stage.
Infrared sensor turns the microscope off after 15 minutes to save power when the "auto" mode is selected. Microscopes also feature a phase slider, coarse and fine focusing system with tension adjustment, and eyepieces inclined at 45°.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Head Configuration Maximum Magnification
Availability Pricing
Motic Instruments 1100103800042 Inverted Microscope, Binocular head WW-48412-10 Mfr # 1100103800042
COMPARE
Binocular 200x
Call For Price
Motic Instruments 1100103800034 Inverted Microscope, Ergonomic Binocular head WW-48412-11 Mfr # 1100103800034
COMPARE
Binocular with ergonomic head 100x
Call For Price
Motic Instruments 1100103800029 Inverted Microscope, Trinocular head WW-48412-12 Mfr # 1100103800029
COMPARE
Trinocular 200x