VistaLab™ MLA™ Micro D-Tipper™ Fixed-Volume Pipettors
Two-stroke operation with overblow feature enhances accuracy and precision at microvolumes
  • Built-in tip ejector
  • Two-stroke operation
  • Color-coded for easy identification
Achieve repeatable and dependable results with your microvolume samples. The MLA™ Micro D-Tipper™ pipettors are ideal for general-purpose use when convenience, accuracy, and precision are required. A simple upward lift on the pipette bonnet ejects the tips with ease. Calibrated “to deliver”, pipettes can be adjusted above or below stated volume ±10%. Easily verify the calibration right in your lab with the calibration key (included).
These fixed-volume pipets are color-coded for easy identification and come with the stated volume engraved onto the piston assembly. Engineered with aluminum alloy and stainless steel, these pipettors withstand years of use and provide exceptional performance for a wide range of fluids, including dense or viscous samples.
55,600.00 - ₹59,100.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy (%) Color Code Max Volume (µL)
Availability Pricing
VistaLab MLA™ Micro D-Tipper™ Fixed-Volume Pipettor, Red, 2 µL WW-06199-45 Mfr # 1802
COMPARE
±2.00% Red 2
₹55,600.00INR / Each
VistaLab MLA™ Micro D-Tipper™ Fixed-Volume Pipettor, Silver, 5 µL WW-06199-51 Mfr # 1805
COMPARE
±1.00% Silver 5
₹59,100.00INR / Each
VistaLab MLA™ Micro D-Tipper™ Fixed-Volume Pipettor, Green, 6 µL WW-06199-53 Mfr # 1806
COMPARE
±1.00% Green 6
₹55,600.00INR / Each
VistaLab MLA™ Micro D-Tipper™ Fixed-Volume Pipettor, Orange, 10 µL WW-06199-61 Mfr # 1810
COMPARE
±1.00% Orange 10
₹55,600.00INR / Each