MLA D-Tipper Fixed-Volume Pipettes
Optimal accuracy and performance for routine tasks
  • Built-in tip ejector eliminates manual tip removal
  • Glove guard ensures user safety and won’t catch or tear during tip removal
  • Each pipete features an engraved serial number for quality control
These pipettes are made specifically for accuracy and precision, with proven reliability—the robust construction comes with a lifetime warranty. The built-in de-tipping mechanism saves you time when removing pipette tips.
All MLA D-Tipper pipettes are engraved with serial numbers to faciliate traceablilty and calibration reporting for easy quality control. These pipettors come factory calibrated and tested at 21.5°C ±2°C using distilled water. Pipette calibration volume can be changed using the calibration key (included).
The body of the pipette is color coded for easy volume identification. Slim tapered design enables access to narrow diameter tubes and is comfortable even for small hands. Smooth one-stroke action yields reproducible results and requires minimal exertion.
55,900.00 - ₹85,000.00INR / Each
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy (%) Color Code Max Volume (µL)
Availability Pricing
VistaLab 1051C Fixed-Volume Pipette, 10 uL, Orange; Each WW-06198-35 Mfr # 1051C
COMPARE
±5.00% Orange 10
₹55,900.00INR / Each
VistaLab 1052C Fixed-Volume Pipette, 20 uL, Black; Each WW-06198-37 Mfr # 1052C
COMPARE
±3.00% Black 20
₹85,000.00INR / Each
VistaLab 1053C Fixed-Volume Pipette, 25 µL, White; Each WW-06198-39 Mfr # 1053C
COMPARE
±3.00% White 25
₹55,900.00INR / Each
VistaLab 1054C Fixed-Volume Pipette, 50 uL, Green; Each WW-06198-41 Mfr # 1054C
COMPARE
±2.00% Green 50
₹59,600.00INR / Each
VistaLab 1055C Fixed-Volume Pipette, 100 uL, Blue; Each WW-06198-43 Mfr # 1055C
COMPARE
±1.00% Blue 100
₹59,600.00INR / Each
VistaLab 1056C Fixed-Volume Pipette, 200 uL, Red, Small Tip; Each WW-06198-45 Mfr # 1056C
COMPARE
±1.00% Red 200
₹59,600.00INR / Each
VistaLab 1057C Fixed-Volume Pipette, 250 uL, Purple; Each WW-06198-49 Mfr # 1057C
COMPARE
±1.00% Purple 250
₹59,600.00INR / Each
VistaLab 1058C Fixed-Volume Pipette, 300 uL, Brown; Each WW-06198-51 Mfr # 1058C
COMPARE
±1.00% Brown 300
₹59,600.00INR / Each
VistaLab 1059C Fixed-Volume Pipette, 500 uL, Yellow; Each WW-06198-53 Mfr # 1059C
COMPARE
±1.00% Yellow 500
₹55,900.00INR / Each
VistaLab 1060C Fixed-Volume Pipette, 1000 uL, Grey; Each WW-06198-55 Mfr # 1060C
COMPARE
±1.00% Gray 1000
₹59,600.00INR / Each
VistaLab 1142C Fixed-Volume Pipette, 15 uL, Silver; Each WW-06198-61 Mfr # 1142C
COMPARE
±5.00% Silver 15
₹55,900.00INR / Each
VistaLab 1146C Fixed-Volume Pipette, 60 uL, Silver; Each WW-06198-69 Mfr # 1146C
COMPARE
±2.00% Silver 60
₹59,600.00INR / Each
VistaLab 1147C Fixed-Volume Pipette, 70 uL, Silver; Each WW-06198-85 Mfr # 1147C
COMPARE
±2.00% Silver 70
₹55,900.00INR / Each
VistaLab 1151C Fixed-Volume Pipette, 120 uL, Silver; Each WW-06198-93 Mfr # 1151C
COMPARE
±1.00% Silver 120
₹55,900.00INR / Each
VistaLab 1152C Fixed-Volume Pipette, 150 uL, Silver; Each WW-06198-95 Mfr # 1152C
COMPARE
±1.00% Silver 150
₹55,900.00INR / Each
VistaLab 1153C Fixed-Volume Pipette, 400 uL, Silver; Each WW-06198-97 Mfr # 1153C
COMPARE
±1.00% Silver 400
₹55,900.00INR / Each
VistaLab 1157C Fixed-Volume Pipette, 750 uL, Silver; Each WW-06199-01 Mfr # 1157C
COMPARE
±1.00% Silver 750
₹55,900.00INR / Each
VistaLab 1162C Fixed-Volume Pipette, 700 uL, Silver; Each WW-06199-07 Mfr # 1162C
COMPARE
±1.00% Silver 700
₹55,900.00INR / Each
VistaLab 1163C Fixed-Volume Pipette, 900 uL, Silver; Each WW-06199-09 Mfr # 1163C
COMPARE
±1.00% Silver 900
₹55,900.00INR / Each
Showing 1- 19 of 19