Mixer Testing Beads
Test your mixer/impeller system for clumping and settling. These highly visible beads are calibrated to simulate particles in your fluid and let you easily visualize mixing patterns. Use different sizes (color-coded) simultaneously to determine effect on different sized particles.
30,000.00 - ₹53,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Mixer Testing Beads, Red Acetate, 4.5 mm Diameter; 1500 Beads/Jar
₹30,000.00INR / Each
COMPARE
Mixer Testing Beads, Yellow Acetate, 3.0 mm Diameter; 2400 Beads/Jar
₹53,000.00INR / Each