Cole-Parmer® Mini Electrophoresis System
Quick and easy, no spacers or clamps required
  • Visualize your run with the viewing area above the gel box
  • Quickly run gels with selectable parameters using the membrane keypad
  • Compact footprint ideal for small laboratories or classrooms
This complete horizontal mini electrophoresis system includes everything needed to get up and running. The system is designed for separating nucleic acids in agarose gel. Selectable parameters include three preset voltages – 35, 50, and 100V, as well as a timer that has a range of 0 to 99 minutes or for continuous use. The safety lid is vented to dissipate heat during the electrophoresis process. The current will only run through the system when the lid is properly in place.
The gel trays feature a dark contrast strip to help visualize wells while loading. In addition, the lines fluoresce under UV and blue light to help track the progress of the sample during the run.
115,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Buffer Solution Required (mL) Power (VAC) Description
Availability Pricing
COMPARE
230 115 Mini electrophoresis system; 115V
₹115,000.00INR / Each
COMPARE
230 230 Mini electrophoresis system; 230V
₹115,000.00INR / Each