Micropump® A-Mount Cavity-Style Pump Heads
Smooth pulseless flow provides precise, leak-free delivery of fluids
  • Cavity-style pump heads are designed to handle higher inlet pressures.
Magnetically driven, precision-geared pump heads provide smooth, accurate, pulseless fluid delivery. The magnetic drive and static seals keep fluids securely inside and potential contaminants out. Chemical resistant materials handle a wide range of materials (due to wetted parts). Pump heads are compact and easy to service. Modular design offers flexibility—configure to your specific needs.
159,000.00 - ₹181,000.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Flow Rate (mL/rev) Internal Bypass Wetted Parts
Availability Pricing
Micropump L21830 A-Mount Cavity Style Pump Head; Bypass, SS, PTFE, PTFE; 0.91 mL/rev WW-07001-40 Mfr # L21830
COMPARE
0.91 Yes 316 SS, PTFE
₹163,000.00INR / Each
Micropump L21832 A-Mount Cavity Style Pump Head; Bypass, PPS, PPS, PTFE; 0.64 mL/rev WW-07001-80 Mfr # L21832
COMPARE
0.64 Yes 316 SS, PPS, PTFE
₹168,000.00INR / Each
Micropump L22092 A-Mount Cavity Style Pump Head; Bypass, SS, PTFE, PTFE; 0.32 mL/rev WW-07002-17 Mfr # L22092
COMPARE
0.32 Yes 316 SS, PTFE
₹160,000.00INR / Each
Micropump L21833 A-Mount Cavity Style Pump Head; Bypass, SS, PTFE, PTFE; 0.64 mL/rev WW-07002-23 Mfr # L21833
COMPARE
0.64 Yes 316 SS, PTFE
₹159,000.00INR / Each
Micropump L21262 A-Mount Cavity Style Pump Head; SS, PEEK, PTFE; 0.32 mL/rev WW-73011-00 Mfr # 85731
COMPARE
0.32 No 316 SS, PEEK, PTFE
₹165,000.00INR / Each
Micropump L21755 A-Mount Cavity Style Pump Head; Bypass, SS, PEEK, PTFE; 0.32 mL/rev WW-73011-02 Mfr # 85702
COMPARE
0.32 Yes 316 SS, PEEK, PTFE
₹170,000.00INR / Each
Micropump L18428 A-Mount Cavity Style Pump Head; SS, PEEK, PTFE; 0.64 mL/rev WW-73011-08 Mfr # 85703
COMPARE
0.64 No 316 SS, PEEK, PTFE
₹161,000.00INR / Each
Micropump L17121 A-Mount Cavity Style Pump Head; Bypass, SS, PEEK, PTFE; 0.64 mL/rev WW-73011-10 Mfr # 85732
COMPARE
0.64 Yes 316 SS, PEEK, PTFE
₹173,000.00INR / Each
Micropump L16516 A-Mount Cavity Style Pump Head; SS, PEEK, PTFE; 0.91 mL/rev WW-73011-16 Mfr # 85711
COMPARE
0.91 No 316 SS, PEEK, PTFE
₹181,000.00INR / Each
Micropump T80 L21267 A-Mount Cavity Style Pump Head; Bypass, SS, PEEK, PTFE; 0.91 mL/rev WW-73011-18 Mfr # 85712
COMPARE
0.91 Yes 316 SS, PEEK, PTFE
₹168,000.00INR / Each
Showing 1- 10 of 10