Microcentrifuge Tube Ice Tray, polypropylene
For temperature stabilized benchtop ice storage
This two piece rack is designed for long life and heavy usage. Rack holds up to fifty 1.5 ml microcentrifuge tubes or other tubes with a diameter under 12 mm. The deep tray holds more ice flakes and water for extended lab work. A flat perimeter rim provides space for labeling tape. Racks may be stacked with identical racks when loaded or empty. All parts are molded of sturdy polypropylene and are autoclavable at 121°C (250°C). An optional polypropylene cover protects tubes during transport or storage.
4,200.00 - ₹12,500.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Description
Availability Pricing
Scienceware H18905-0001 Microcentrifuge Tube Rack (holds 50 X 1.5 mL Tubes) WW-06274-07 Mfr # H18905-0001
COMPARE
Polypropylene(PP) Microcentrifuge Tube Rack (holds 50 X 1.5 mL Tubes)
₹12,500.00INR / Each
Scienceware F18905-0011 Microcentrifuge Tube Rack Cover, polypropylene WW-06274-08 Mfr # F18905-0011
COMPARE
Polypropylene(PP) Microcentrifuge Tube Rack Cover, polypropylene
₹4,200.00INR / Each