Mettler Toledo® Remote / Column / Auxiliary Displays
Add an extra balance display to create a truly ergonomic workspace
This auxiliary LCD display with backlighting can be placed up to 10 meters from the balance due to the wireless Bluetooth connection. The extra display can be placed elsewhere on your lab bench, outside a fume cupboard or mounted on a stand or wall.
141,000.00 - ₹154,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Accessory Type Height (in) Length (in)
Availability Pricing
Mettler Toledo 12122381 Auxiliary Display For RS232/PL-E Balances WW-11140-30 Mfr # 12122381
COMPARE
₹141,000.00INR / Each
Mettler Toledo 30125077 Ergostand For XPE S-Platforms WW-11140-89 Mfr # 30125077
COMPARE
19 1/4
₹154,000.00INR / Each