Mettler Toledo Portable Refractometers
Reliable results within seconds
  • Ensure accuracy with the built-in temperature compensation
  • Quickly view Brix or refractive index measurements with the click of a button
  • Easily transfer important data to a PC or printer for documentation
The Mettler Toledo 30PX and 30GS portable refractometers are suited for determining the refractive index of liquids, providing temperature-compensated results. These are handheld and benchtop refractometers in one, and the cell can be immersed directly in the sample or a sample added to the cell for increased flexibility. Both options allow you to save 1100 results – including information such as sample and instrument identification, measurement unit, temperature correction coefficient, date, and time. Both units feature a backlit LCD for easy-to-read measurements, and can be interfaced to a PC or printer for exporting data.
929,000.00 - ₹1,256,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Brix Range (%) Range Description
Availability Pricing
Mettler Toledo Refracto 30PX Portable Handheld Refractometer WW-33333-37 Mfr # 51324650
COMPARE
85 0 to 85% Brix, 1.32 to 1.50 RI Mettler Toledo Refracto 30PX Portable Handheld Refractometer
₹929,000.00INR / Each
Mettler Toledo Refracto 30GS Portable Handheld Refractometer WW-33333-38 Mfr # 51324660
COMPARE
85 0 to 85% Brix, 1.32 to 1.65 RI Mettler Toledo Refracto 30GS Portable Handheld Refractometer
₹1,256,000.00INR / Each