Mettler Toledo NTEP Approved Excellence XPR Precision Toploading Balances
Built-in quality assurance monitoring ensures data integrity
  • NTEP approved legal for trade
  • StatusLight™ uses colored lights on terminal to indicate when balance is ready for weighing
  • Performs temperature-activated internal calibrations automatically at preprogrammed intervals
  • Integrated LabX® software automatically saves all data results and provides high process security
  • SmartPan™ weighing pan reduces effect of turbulence, shortening stabilization time and repeatability
  • Graphical leveling device provides warning when balance is not level
  • Overload protection safeguards weighing cell against damage
  • Built-in notepad automatically records all task parameters and results
  • 7-inch color touch screen provides clear visibility of weighing results—is glove compatible
  • Password-protected individual profiles
The XPR series balances feature the GWP approved built-in quality assurance functions which automatically monitor the balance level, routine tests, and minimum weight limit, ensuring your results are always valid and comply with the strictest regulations. State-of-the-art user interface with intelligent functions helps simplify and supports your processes at every step. |The innovative SmartPan weighing pan minimizes air currents on the weighing cell, improving stabilization time and repeatability, allowing you to get results twice as fast as standard weighing pans without the use of a draft shield. |An intuitive touch screen streamlines operations, provides quick access to menus and tasks, and allows you to set up shortcuts, and use multiple fingers to enter data. Store regularly used task methods in the method library for quick future recall. All task parameters are recorded on the results notepad where you can add your own comments. Easily transfer data to a PC, printer, or bar code reader via four USB or Ethernet ports. No more manual transcriptions. |The balances are designed with chemical-resistant materials, smooth surfaces, and rounded edges, making them easy to clean.
2,467,000.00 - ₹3,464,000.00INR / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (mg)
Availability Pricing
Mettler Toledo XPR12001L/A Excellence XPR Precisions Legal for Trade Toploader Balance, Large, 12.1kg x, 0.1g, Touchscreen WW-11338-01 Mfr # XPR12001L/A
COMPARE
Internal 10100 100
Call For Price
Mettler Toledo XPR64000L/A Excellence XPR Precisions Legal for Trade Toploader Balance, Large, 64.1kg x, 1g, Touchscreen WW-11338-11 Mfr # XPR64000L/A
COMPARE
Internal 64100 1000
Call For Price
Mettler Toledo XPR15002L/A Excellence XPR Precisions Legal for Trade Toploader Balance, Large, 15.1kg x, 0.01g, SmartPan Weighing Pan, Touchscreen WW-11338-13 Mfr # XPR15002L/A
COMPARE
Internal 15100 10
Call For Price
Mettler Toledo XPR504S/A Excellence XPR Precisions Legal for Trade Toploader Balance, 510g x, 0.1mg, SmartPan Weighing Pan, Touchscreen WW-11338-21 Mfr # XPR504S/A
COMPARE
Internal 510 0.1
Call For Price
Mettler Toledo XPR303SN/A Excellence Precision XPR Legal for Trade Toploader Balance, 310g x, 1mg, No Draft Shield, SmartPan Weighing Pan, Touchscreen WW-11338-27 Mfr # XPR303SN/A
COMPARE
Internal 310 1
Call For Price
Mettler Toledo XPR4002S/A Excellence Precision XPR Legal for Trade Toploader Balance, 4.1kg x, 0.01g, SmartPan Weighing Pan, Touchscreen WW-11338-47 Mfr # XPR4002S/A
COMPARE
Internal 4100 10
Call For Price
Mettler Toledo XPR6002S/A Excellence Precision XPR Legal for Trade Toploader Balance, 6.1kg x, 0.01g, SmartPan Weighing Pan, Touchscreen WW-11338-49 Mfr # XPR6002S/A
COMPARE
Internal 6100 10
Call For Price
Mettler Toledo XPR1203SN/A Excellence XPR Precision Legal for Trade Toploader Balance, Large , 1201g x , 0.001g , Touchscreen WW-11348-96 Mfr # XPR1203SN/A
COMPARE
Internal 1210 1
₹2,741,000.00INR / Each
Mettler Toledo XPR2003S/A Excellence XPR Precision Legal for Trade Toploader Balance, Large , 2100g x , 0.001g , Touchscreen WW-11348-98 Mfr # XPR2003S/A
COMPARE
Internal 2100 1
₹2,900,000.00INR / Each
Mettler Toledo XPR12002S/A Excellence XPR Precision Legal for Trade Toploader Balance, Large , 12100g x , 0.01g , Touchscreen WW-11349-01 Mfr # XPR12002S/A
COMPARE
Internal 12100 10
₹3,464,000.00INR / Each
Mettler Toledo XPR12001S/A Excellence XPR Precision Legal for Trade Toploader Balance, Large , 12100g x , 0.1g , Touchscreen WW-11349-04 Mfr # XPR12001S/A
COMPARE
Internal 12100 100
₹2,467,000.00INR / Each
Showing 1- 11 of 11