Mettler Toledo NewClassic ME-T Analytical Legal for Trade (NTEP) Balances
Worry-free weighing with NTEP certification provides confidence that a device will be manufactured in accordance with United States standards
  • Large color touchscreen and intuitive interface simplify operation
  • Fully Automatic Calibration Technology (FACT) ensures accuracy
  • Eight embedded applications include essential weighing functions, statistics, and formulation
  • Sample ID and passcode protection produce GLP documentation of results
  • RS-232 and USB connectivity simplify data upload and transfer
Analytical NTEP balances feature a 4.5” color TFT touchscreen with intuitive guidance enabling you to operate quickly and efficiently, even while wearing gloves. An electromagnetic force compensation (EMFC) weighing cell with fully automated calibration technology (FACT) automatically adjusts the balance to provide consistent reliable results. Multiple interfaces support barcode reader, printer and PC connections for easy traceability and data handling. USB and RS-232 connectivity along with an optional Bluetooth adapter (sold separately) simplify data upload and transfer.
994,000.00 - ₹1,104,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (g)
Availability Pricing
Mettler Toledo NewClassic ME-T Legal for Trade (NTEP) Analytical Balance, 120 g x 0.1 mg, Internal Calibration WW-11348-14 Mfr # ME104T/A
COMPARE
120 0.0001
₹994,000.00INR / Each
Mettler Toledo NewClassic ME-T Legal for Trade (NTEP) Analytical Balance, 220 g x 0.1 mg, Internal Calibration WW-11348-15 Mfr # ME204T/A
COMPARE
220 0.0001
₹1,104,000.00INR / Each