Mettler Toledo NewClassic ME-T Analytical Balances, Internal Calibration
Easy worry-free weighing with accurate, reliable results day after day
  • Large color touchscreen and intuitive interface to simplify operation
  • Ten embedded applications include essential weighing functions, statistics, and formulation
  • Electromagnetic Force Compensation (EMFC) ensures accuracy
  • Sample ID and passcode protection produce GLP documentation of results
  • Built-in connectivity includes RS-232, USB host, and USB device
The ME-T analytical balance is great for your sensitive, yet routine weighing procedures. The bright 4.5” color TFT touchscreen with intuitive guidance enables you to operate quickly and efficiently, even when wearing gloves. The multilingual text and graphics walk you through each step.
An electromagnetic force compensation (EMFC) weighing cell with fully automated calibration technology (FACT) automatically adjusts the balance to compensate for a change in ambient temperature, to provide consistent reliable results. Built-in overload protection prolongs the life of your balance.
Multiple interfaces support a barcode reader, printer, and PC connections for easy traceability and data handling.
885,000.00 - ₹1,063,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (mg)
Availability Pricing
Mettler Toledo ME54T NewClassic ME-T Standard Analytical Balance, 52 g x 0.1 mg WW-11348-00 Mfr # ME54T
COMPARE
Internal 52 0.1
₹885,000.00INR / Each
Mettler Toledo ME104T NewClassic ME-T Standard Analytical Balance, 120 g x 0.1 mg WW-11348-01 Mfr # ME104T
COMPARE
Internal 120 0.1
₹956,000.00INR / Each
Mettler Toledo ME204T NewClassic ME-T Standard Analytical Balance, 220 g x 0.1 mg WW-11348-02 Mfr # ME204T
COMPARE
Internal 220 0.1
₹1,063,000.00INR / Each