METTLER TOLEDO Laboratory Dissolved Oxygen Probes
Robust construction for harsh environments
  • IP67-rated to withstand dirty, wet conditions
  • Durable plastic housing protects sensor
  • Intelligent Sensor Management (ISM) ensures accurate, repeatable measurements in the field
Choose between polarographic and Optical RDO.
237,000.00 - ₹281,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Mettler Toledo 605 Dissolved Oxygen Probe with ISM; 1.8 m cable WW-35903-01 Mfr # 51344611
COMPARE
Dissolved Oxygen Probe with ISM; 1.8 m cable
₹237,000.00INR / Each
Mettler Toledo OptiOx Optical Rugged Dissolved Oxygen Probe with ISM; 1.8 m cable WW-35903-07 Mfr # 51344621
COMPARE
Optical Rugged Dissolved Oxygen Probe with ISM; 1.8 m cable
₹281,000.00INR / Each