Mettler Toledo LA Series Affordable Quality Analytical Balances
Quality balances offer accurate and reliable results at a low cost
  • Electromagnetic Force Compensation (EMFC) weighing cell provides accurate and reliable results
  • Easy-to-read, bright backlit LCD
  • Connect to a printer, secondary display, or computer via the RS-232 interface
  • Make adjustments with external weights to ensure accuracy
  • Overload protection guards the weighing cell against excess weight overload
  • Durable die-cast aluminum base and ABS top housing
The LA series analytical balances provide reliable performance at a low cost. Balances are simple to use yet feature three built-in applications to perform calculations automatically, ensuring consistent processes and fewer errors. Perform weighing operations below the balance with the included weighing hook.
Rugged housing, made of high-quality and durable materials, protects the sensitive sensor from environmental influences and impacts. Setting protection feature protects balance configurations against unintended changes. Built-in overload protection prolongs the life of your balance.
522,000.00 - ₹584,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (mg)
Availability Pricing
Mettler Toledo LA84E Analytical Balance, LA Series, 82g x 0.1mg with LCD Display and External Calibration WW-02629-61 Mfr # 30823718
COMPARE
External 82 0.1
₹522,000.00INR / Each
Mettler Toledo LA104E Analytical Balance, LA Series, 120g x 0.1mg with LCD Display and External Calibration WW-02629-62 Mfr # 30823719
COMPARE
External 120 0.1
₹543,000.00INR / Each
Mettler Toledo LA204E Analytical Balance, LA Series, 220g x 0.1mg with LCD Display and External Calibration WW-02629-63 Mfr # 30823720
COMPARE
External 220 0.1
₹584,000.00INR / Each