Mettler Toledo Excellence XSR Precision Toploading Balances
Delivers fast, accurate results under the most challenging environments
  • MonoBloc™ weighing cell offers fast, precise results
  • StatusLight™ uses colored lights on terminal to indicate when balance is ready for weighing
  • Performs temperature-activated internal calibrations automatically at preprogrammed intervals
  • Optional LabX® software automatically saves all data results and provides high process security
  • Graphical leveling device provides warning when balance is not level
  • Overload protection safeguards weighing cell against damage
  • Built-in notepad automatically records all task parameters and results
  • 4.3-inch color touch screen provides clear visibility of weighing results—is glove compatible
  • Password-protected configuration
The XSR series balances are ideal for handling high sample throughput with tight timelines even under challenging production environments. State-of-the-art user interface with intelligent functions simplifies and supports your processes at every step, while the MonoBloc™ high-speed weighing cell delivers fast, precise results.
The innovative SmartPan weighing pan minimizes air currents on the weighing cell, improving stabilization time and repeatability, allowing you to get results twice as fast as standard weighing pans without the use of a draft shield.
An intuitive touch screen streamlines operations, provides quick access to menus and tasks, and allows you to set up shortcuts and use multiple fingers to enter data. Store regularly used task methods in the method library for future recall. All task parameters are recorded on the results notepad where you can add your own comments. Easily transfer data to a PC, printer, or bar code reader via four USB or Ethernet ports. No more manual transcriptions.
The balances are designed with chemical-resistant materials, smooth surfaces, and rounded edges making them easy to clean.
Call For Price
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
Mettler Toledo XSR10001L Excellence XSR Precision Toploader Balance, Large Solutions, 10.1kg x 0.1g, SmartPan Weighing Pan, Touchscreen WW-11338-65 Mfr # XSR10001L
COMPARE
Internal 10100 0.1
Call For Price
Mettler Toledo XSR16001L Excellence XSR Precision Toploader Balance, Large Solutions, 16.1kg x 0.1g, Touchscreen WW-11338-67 Mfr # XSR16001L
COMPARE
Internal 16100 0.1
Call For Price
Mettler Toledo XSR32001L Excellence XSR Precisions Toploader Balance, 32.1kg x 0.1g, Touchscreen WW-11338-69 Mfr # XSR32001L
COMPARE
Internal 32100 0.1
Call For Price
Mettler Toledo XSR32001LDR Excellence XSR Precision Toploader Balance, Large, 32.1kg /6.4kg x 1g /0.1g, Touchscreen WW-11338-71 Mfr # XSR32001LDR
COMPARE
Internal 32100 1
Call For Price
Mettler Toledo XSR32000L Excellence XSR Precision Toploader Balance, Large, 32.1kg x 1g, SmartPan Weighing Pan, Touchscreen WW-11338-75 Mfr # XSR32000L
COMPARE
Internal 32100 1
Call For Price
Mettler Toledo XSR303SN Excellence Precision XSR Toploader Balance, 310g x 1mg, No Draft Shield, SmartPan Weighing Pan, Touchscreen WW-11338-79 Mfr # XSR303SN
COMPARE
Internal 310 1
Call For Price
Mettler Toledo XSR603S Excellence XSR Precision Toploader Balance, 610g x 1mg, SmartPan Weighing Pan, Touchscreen WW-11338-81 Mfr # XSR603S
COMPARE
Internal 610 1
Call For Price
Mettler Toledo XSR1203S Excellence XSR Precision Toploader Balance, 1,210g x 1mg, SmartPan Weighing Pan, Touchscreen WW-11338-85 Mfr # XSR1203S
COMPARE
Internal 1210 1
Call For Price
Mettler Toledo XSR2002S Excellence Precision XSR Toploader Balance, 2,100g x 0.01g, SmartPan Weighing Pan, Touchscreen WW-11338-89 Mfr # XSR2002S
COMPARE
Internal 2100 0.01
Call For Price
Mettler Toledo XSR4002S Excellence XSR Precisions Toploader Balance, 4.1kg x 0.01g, SmartPan Weighing Pan, Touchscreen WW-11338-91 Mfr # XSR4002S
COMPARE
Internal 4100 0.01
Call For Price
Mettler Toledo XSR6002S Excellence Precision XSR Toploader Balance, 6.1kg x 0.01g, SmartPan Weighing Pan, Touchscreen WW-11338-93 Mfr # XSR6002S
COMPARE
Internal 6100 0.01
Call For Price
Mettler Toledo XSR6002SDR Excellence XSR Precision Toploader Balance, 6.1kg /1.2kg x 0.1g /0.01g, SmartPan Weighing Pan, Touchscreen WW-11338-95 Mfr # XSR6002SDR
COMPARE
Internal 64100 0.01
Call For Price
Mettler Toledo XSR6001S Excellence XSR Precision Toploader Balance, 6.1kg x 0.1g, SmartPan Weighing Pan, Touchscreen WW-11339-01 Mfr # XSR6001S
COMPARE
Internal 6100 0.1
Call For Price
Mettler Toledo XSR8001S Excellence XSR Precision Toploader Balance, 8.1kg x 0.1g, SmartPan Weighing Pan, Touchscreen WW-11339-03 Mfr # XSR8001S
COMPARE
Internal 8100 0.1
Call For Price
Mettler Toledo XSR10001S Excellence Precision XSR Toploader Balance, 10.1kg x 0.1g, SmartPan Weighin Pan, Touchscreen WW-11339-05 Mfr # XSR10001S
COMPARE
Internal 10100 0.1
Call For Price
Showing 1- 15 of 15