This product is now discontinued, View Related Products
Mettler Toledo Benchtop Digital Refractometer
Reliable results within seconds
  • Ensure accuracy with the built-in temperature compensation
  • Quickly view Brix or refractive index measurements with the click of a button
  • Easily transfer important data to a PC or printer for documentation
The Mettler Toledo RM40 and RM50 benchtop refractometers are compact, easy-to-use units suitable for a vast diversity of applications. They have extremely fast heating up-and cooling down rates, maximum accuracy in a minimum amount of time and are simple to clean – the top section can be easily removed and rinsed with tap water without the need for any tools. The color touch screen allows for measurement of your sample with only one click. These feature automated QC results for each product with user-defined upper and lower limits. Both units have a measurement duration time of approximately 15 seconds for quick and accurate results and a minimum sample volume of 0.5 mL. These allow for up to 30 individual users to be assigned and password protected. Both models have a built-in Peltier thermostat and can be connected to a printer, computer, USB stick, or other external devices for easy data handling.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Brix Range (%) Range Description
Availability Pricing
Mettler Toledo RM40 LiquiPhysics Excellence Refractometer, nD 0.00005 WW-33333-39 Mfr # 51337003
COMPARE
85 0 to 85% Brix, 1.3200 to 1.7000 RI Mettler Toledo RM40 LiquiPhysics Excellence Refractometer, nD 0.00005
Call For Price
Mettler Toledo RM50 LiquiPhysics Excellence Refractometer, nD 0.00001 WW-33333-40 Mfr # 51337004
COMPARE
85 0 to 85% Brix, 1.32000 to 1.58000 RI Mettler Toledo RM50 LiquiPhysics Excellence Refractometer, nD 0.00001