This product is now discontinued, View Related Products

Meriam Ruggedized Well-Type Manometers

Accurate and dependable, as well as highly versatile
  • Unit scale extends the full width of the instrument body and is graduated on each side of the indicating column
  • Scale has a white, nonglare background with black graduations and figures
  • Table- or wall-mount style
Well-type manometers provide fast, direct readings of pressure, vacuum, differential, flow and liquid levels. No adding of distances above and below a zero line. All scales are graduated
to correct for the drop in well level and are full width of the instrument.

Ranges: 6, 12, 20, 30, 40, and 60 inches
Call For Price

50 variations of this product are available.

See All Product Options Below
50 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
Meriam 923RO-1 Red Oil for Manometers, 0.827 SG, 4 fl oz WW-68460-04 Mfr # 923RO-1
COMPARE
Call For Price
Meriam 924BA-1 Red Fluid, 2.95SP.Gr Manometer Fluid 1LB. WW-68460-06 Mfr # 924BA-1
COMPARE
Call For Price
Meriam 30EBX25-012-01-1-02 Ruggedized Well-Type Wall-Mount Manometers, Scale A, 12" WC WW-68460-07 Mfr # 30EBX25-012-01-1-02
COMPARE
Call For Price
Meriam 30EBX25-020-01-1-02 Ruggedized Well-Type Wall-Mount Manometers, Scale A, 20" WC WW-68460-08 Mfr # 30EBX25-020-01-1-02
COMPARE
Call For Price
Meriam 30EBX25-030-01-1-02 Ruggedized Well-Type Wall-Mount Manometers, Scale A, 30" WC WW-68460-09 Mfr # 30EBX25-030-01-1-02
COMPARE
Call For Price
Meriam 30EBX25-040-01-1-02 Ruggedized Well-Type Wall-Mount Manometers, Scale A, 40" WC WW-68460-10 Mfr # 30EBX25-040-01-1-02
COMPARE
Call For Price
Meriam 30EBX25-060-01-1-02 Ruggedized Well-Type Wall-Mount Manometers, Scale A, 60" WC WW-68460-11 Mfr # 30EBX25-060-01-1-02
COMPARE
Call For Price
Meriam 30EBX25-020-01-2-02 Ruggedized Well-Type Table-Mount Manometers, Scale A, 20" WC WW-68460-12 Mfr # 30EBX25-020-01-2-02
COMPARE
±0.5% of smallest graduation
Call For Price
Meriam 30EBX25-030-01-2-02 Ruggedized Well-Type Table-Mount Manometers, Scale A, 30" WC WW-68460-13 Mfr # 30EBX25-030-01-2-02
COMPARE
±0.5% of smallest graduation
Call For Price
Meriam 30EBX25-040-01-2-02 Ruggedized Well-Type Table-Mount Manometers, Scale A, 40" WC WW-68460-14 Mfr # 30EBX25-040-01-2-02
COMPARE
±0.5% of smallest graduation
Call For Price
Meriam 30EBX25-060-01-2-02 Ruggedized Well-Type Table-Mount Manometers, Scale A, 60" WC WW-68460-15 Mfr # 30EBX25-060-01-2-02
COMPARE
±0.5% of smallest graduation
Call For Price
Meriam 30EBX25-6-03-1 Ruggedized Well-Type Wall-Mount Manometers, Scale G, 4.9" WC WW-68500-31 Mfr # 30EBX25-6-03-1
COMPARE
Call For Price
Meriam 30EBX25-12-03-1 Ruggedized Well-Type Wall-Mount Manometers, Scale G, 9.9" WC WW-68500-32 Mfr # 30EBX25-12-03-1
COMPARE
Call For Price
Meriam 30EBX25-20-03-1 Ruggedized Well-Type Wall-Mount Manometers, Scale G, 16.5" WC WW-68500-33 Mfr # 30EBX25-20-03-1
COMPARE
Call For Price
Meriam 30EBX25-30-03-1 Ruggedized Well-Type Wall-Mount Manometers, Scale G, 24.7" WC WW-68500-34 Mfr # 30EBX25-30-03-1
COMPARE
Call For Price
Meriam 30EBX25-40-03-1 Ruggedized Well-Type Wall-Mount Manometers, Scale G, 33" WC WW-68500-35 Mfr # 30EBX25-40-03-1
COMPARE
Call For Price
Meriam 30EBX25-60-03-1 Ruggedized Well-Type Wall-Mount Manometers, Scale G, 49.5" WC WW-68500-36 Mfr # 30EBX25-60-03-1
COMPARE
Call For Price
Meriam 30EBX25-6-03-2 Ruggedized Well-Type Table-Mount Manometers, Scale G, 4.9" WC WW-68500-37 Mfr # 30EBX25-6-03-2
COMPARE
Call For Price
Meriam 30EBX25-12-03-2 Ruggedized Well-Type Table-Mount Manometers, Scale G, 9.9" WC WW-68500-38 Mfr # 30EBX25-12-03-2
COMPARE
Call For Price
Meriam 30EBX25-20-03-2 Ruggedized Well-Type Table-Mount Manometers, Scale G, 16.5" WC WW-68500-39 Mfr # 30EBX25-20-03-2
COMPARE
Call For Price
Showing 1 - 20 of 50

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat