This product is now discontinued, View Related Products
Memosens Digital Sensors for Knick Meters
Plug-and-play digital sensor technology for pH, conductivity and dissolved oxygen
  • Add additional parameters by simply adding optional Memosens sensors
The Memosens sensors digitally store their serial number, electrode type, unique ID, and calibration data. Easily change your meter from DO to pH to conductivity with Memosens smart sensor recognition. The sensors can also be moved from one meter to another and transfer their calibration data.
Call For Price
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Calibration Temperature compensation Waterproof
Availability Pricing
Memosens CA/MS-003NAA Cables, 3 M ( Digital Sensors for Knick Meters) WW-56030-32 Mfr # CA/MS-003NAA
COMPARE
Call For Price
Memosens CA/MS-005NAA Cables, 5 M ( Digital Sensors for Knick Meters) WW-56030-34 Mfr # CA/MS-005NAA
COMPARE
Call For Price
Memosens CA/MS-010NAA Cables, 10 M ( Digital Sensors for Knick Meters) WW-56030-36 Mfr # CA/MS-010NAA
COMPARE
Call For Price
Memosens SE533X/1-NMSN Digital Sensor, High Pressure pH Electrode WW-56030-51 Mfr # SE533X/1-NMSN
COMPARE
Call For Price
Memosens SE559X/1-NMSN Digital Sensor, Gel pH Electrode WW-56030-53 Mfr # SE559X/1-NMSN
COMPARE
Call For Price
Memosens SE554X/1-NMSN Stratos Digital Sensor, Ultra-Pure Water pH Electrode WW-56030-57 Mfr # SE554X/1-NMSN
COMPARE
Call For Price
COMPARE
Call For Price
COMPARE
Call For Price
Memosens pH Sensor Replacement Digital pH Electrode WW-59351-06 Mfr # SE101NMS
COMPARE
Call For Price
Memosens SE604X-MS Replacement Conductivity Sensor/Probe WW-59351-10 Mfr # SE604X-MS
COMPARE
Call For Price
Memosens Optical DO Replacement Stratos Probe for Knick Meters WW-59351-16 Mfr # SE340
COMPARE
Call For Price
Memosens Optical DO Cap Stratos Replacement DO Sensor Cap WW-59351-18 Mfr # ZU0913
COMPARE
Call For Price
Showing 1- 12 of 12